Assay Details
| Gene Symbol: LOC4335865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780926.2 | XP_015636412.1 | 1 - 2 | 116 | 90 | |
| GenBank mRNA | AK102190.1 | - | 1 - 2 | 99 | 90 | |
| BT130963.1 | - | 1 - 2 | 92 | 90 | ||
| BT132033.1 | - | 1 - 2 | 97 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780926.2 transcripts.
Set Membership: |
|

LOC4335865
Genomic Map