Assay Details
Gene Symbol: LOC9271714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780375.2 | XP_015635861.1 | 11 - 12 | 1380 | 118 | |
XM_015780376.2 | XP_015635862.1 | 9 - 10 | 620 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
