Assay Details
Gene Symbol: LOC9271714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780375.2 | XP_015635861.1 | 13 - 14 | 1586 | 110 | |
XM_015780376.2 | XP_015635862.1 | 11 - 12 | 826 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
