Assay Details
| Gene Symbol: LOC4336039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778757.2 | XP_015634243.1 | 1 - 2 | 1348 | 63 | |
| GenBank mRNA | AK106058.1 | - | 1 - 2 | 254 | 63 | |
More Information
Set Membership: |
|

LOC4336039
Genomic Map