Assay Details
| Gene Symbol: LOC4336129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778632.2 | XP_015634118.1 | 3 - 4 | 633 | 132 | |
| GenBank mRNA | AK070339.1 | - | 3 - 4 | 634 | 132 | |
More Information
Set Membership: |
|

LOC4336129
Genomic Map