Assay Details
| Gene Symbol: LOC4336129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778632.2 | XP_015634118.1 | 8 - 9 | 2083 | 57 | |
| GenBank mRNA | AK070336.1 | - | 5 - 6 | 1059 | 57 | |
| AK070339.1 | - | 8 - 9 | 2084 | 57 | ||
More Information
Set Membership: |
|

LOC4336129
Genomic Map