Assay Details
| Gene Symbol: LOC4336257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780844.1 | XP_015636330.1 | 31 - 32 | 3788 | 154 | |
| XM_015780845.2 | XP_015636331.1 | 29 - 30 | 4009 | 154 | ||
| XM_015780846.1 | XP_015636332.1 | 30 - 31 | 3734 | 154 | ||
| XM_015780847.1 | XP_015636333.1 | 31 - 32 | 3707 | 154 | ||
| GenBank mRNA | AK072226.1 | - | 13 - 14 | 1330 | 154 | |
More Information
Set Membership: |
|

LOC4336257
Genomic Map