Assay Details
| Gene Symbol: LOC4336257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780844.1 | XP_015636330.1 | 26 - 27 | 3228 | 79 | |
| XM_015780845.2 | XP_015636331.1 | 24 - 25 | 3449 | 79 | ||
| XM_015780846.1 | XP_015636332.1 | 25 - 26 | 3174 | 79 | ||
| XM_015780847.1 | XP_015636333.1 | 26 - 27 | 3147 | 79 | ||
| GenBank mRNA | AK072226.1 | - | 8 - 9 | 770 | 79 | |
More Information
Set Membership: |
|

LOC4336257
Genomic Map