Assay Details
Gene Symbol: LOC4336462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780588.2 | XP_015636074.1 | 2 - 3 | 310 | 67 | |
GenBank mRNA | AK072980.1 | - | 2 - 3 | 258 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
