Assay Details
Gene Symbol: LOC4336500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780748.2 | XP_015636234.1 | 2 - 3 | 755 | 65 | |
GenBank mRNA | AK107135.1 | - | 2 - 3 | 455 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
