Assay Details
| Gene Symbol: LOC4337061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779130.2 | 4 - 5 | 1025 | 95 | ||
| GenBank mRNA | AK062930.1 | - | 3 - 4 | 324 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4337061| Gene Symbol: LOC4337061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779130.2 | 4 - 5 | 1025 | 95 | ||
| GenBank mRNA | AK062930.1 | - | 3 - 4 | 324 | 95 | |
Set Membership: |
|