Assay Details
Gene Symbol: LOC4338728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784971.2 | XP_015640457.1 | 2 - 3 | 1297 | 121 | |
GenBank mRNA | AK240941.1 | - | 2 - 3 | 527 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
