Assay Details
Gene Symbol: LOC4338728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784971.2 | XP_015640457.1 | 3 - 4 | 1462 | 76 | |
GenBank mRNA | AK240941.1 | - | 3 - 4 | 692 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
