Assay Details
Gene Symbol: LOC4338736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785064.2 | XP_015640550.1 | 3 - 4 | 672 | 81 | |
GenBank mRNA | AK070043.1 | - | 3 - 4 | 660 | 81 | |
AK104199.1 | - | 3 - 4 | 659 | 81 | ||
AK104366.1 | - | 3 - 4 | 659 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
