Assay Details
Gene Symbol: LOC4339821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783930.2 | XP_015639416.1 | 11 - 12 | 1600 | 66 | |
GenBank mRNA | AK121149.1 | - | 11 - 12 | 1546 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
