Assay Details
Gene Symbol: LOC4339821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783930.2 | XP_015639416.1 | 16 - 17 | 2195 | 138 | |
GenBank mRNA | AK121149.1 | - | 16 - 17 | 2141 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
