Assay Details
Gene Symbol: LOC4339821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783930.2 | XP_015639416.1 | 22 - 23 | 2779 | 125 | |
GenBank mRNA | AK121149.1 | - | 22 - 23 | 2725 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
