Assay Details
Gene Symbol: LOC4339821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783930.2 | XP_015639416.1 | 24 - 25 | 2931 | 109 | |
GenBank mRNA | AK121149.1 | - | 24 - 25 | 2877 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
