Assay Details
Gene Symbol: LOC4339821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783930.2 | XP_015639416.1 | 4 - 5 | 739 | 101 | |
GenBank mRNA | AK107823.1 | - | 4 - 5 | 684 | 101 | |
AK121149.1 | - | 4 - 5 | 685 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
