Assay Details
Gene Symbol: LOC4340889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788772.2 | XP_015644258.1 | 7 - 8 | 1516 | 130 | |
GenBank mRNA | AK067233.1 | - | 7 - 8 | 1479 | 130 | |
AK101600.1 | - | 7 - 8 | 1495 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
