Assay Details
Gene Symbol: LOC4340889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788772.2 | XP_015644258.1 | 8 - 9 | 1663 | 72 | |
GenBank mRNA | AK067233.1 | - | 8 - 9 | 1626 | 72 | |
AK101600.1 | - | 8 - 9 | 1642 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
