Assay Details
| Gene Symbol: LOC4341317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785555.2 | XP_015641041.1 | 3 - 4 | 717 | 59 | |
| GenBank mRNA | AK243139.1 | - | 3 - 4 | 518 | 59 | |
More Information
Set Membership: |
|

LOC4341317
Genomic Map