Assay Details
Gene Symbol: LOC4345817 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793625.2 | XP_015649111.1 | 1 | 1086 | 96 | |
GenBank mRNA | AK111147.1 | - | 1 | 947 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
