Assay Details
Gene Symbol: LOC9272626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792804.2 | XP_015648290.1 | 2 - 3 | 588 | 88 | |
GenBank mRNA | AK065590.1 | - | 1 - 2 | 1204 | 88 | |
AK104597.1 | - | 2 - 3 | 566 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
