Assay Details
Gene Symbol: LOC4345945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794953.2 | XP_015650439.1 | 13 - 14 | 2154 | 66 | |
XM_015794954.2 | XP_015650440.1 | 12 - 13 | 2112 | 66 | ||
XM_015794955.1 | XP_015650441.1 | 11 - 12 | 1940 | 66 | ||
GenBank mRNA | AK064832.1 | - | 11 - 12 | 1908 | 66 | |
AK065120.1 | - | 12 - 13 | 2075 | 66 | ||
AY772415.1 | - | 12 - 13 | 2061 | 66 | ||
AY772416.1 | - | 11 - 12 | 1890 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
