Assay Details
Gene Symbol: LOC4345970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795444.2 | XP_015650930.1 | 10 - 11 | 1125 | 82 | |
XM_015795445.2 | XP_015650931.1 | 11 - 12 | 1023 | 82 | ||
GenBank mRNA | AK069190.1 | - | 11 - 12 | 968 | 82 | |
AK072089.1 | - | 10 - 11 | 1108 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
