Assay Details
Gene Symbol: LOC4345970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795444.2 | XP_015650930.1 | 1 - 2 | 184 | 62 | |
XM_015795445.2 | XP_015650931.1 | 1 - 2 | 184 | 62 | ||
GenBank mRNA | AK069190.1 | - | 1 - 2 | 131 | 62 | |
AK072089.1 | - | 1 - 2 | 167 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
