Assay Details
Gene Symbol: LOC4345970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795444.2 | XP_015650930.1 | 2 - 3 | 295 | 72 | |
XM_015795445.2 | XP_015650931.1 | 2 - 3 | 295 | 72 | ||
GenBank mRNA | AK069190.1 | - | 2 - 3 | 242 | 72 | |
AK072089.1 | - | 2 - 3 | 278 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
