Assay Details
Gene Symbol: LOC4345970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795444.2 | XP_015650930.1 | 7 - 8 | 683 | 131 | |
XM_015795445.2 | XP_015650931.1 | 7 - 8 | 683 | 131 | ||
GenBank mRNA | AK069190.1 | - | 7 - 8 | 630 | 131 | |
AK072089.1 | - | 7 - 8 | 666 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
