Assay Details
Gene Symbol: LOC4345970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795445.2 | XP_015650931.1 | 9 - 10 | 846 | 89 | |
GenBank mRNA | AK069190.1 | - | 9 - 10 | 793 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
