Assay Details
| Gene Symbol: LOC4346812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755207.2 | XP_015610693.1 | 4 - 5 | 410 | 98 | |
| GenBank mRNA | AK101995.1 | - | 4 - 5 | 349 | 98 | |
More Information
Set Membership: |
|

LOC4346812
Genomic Map