Assay Details
Gene Symbol: LOC4347697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756538.2 | XP_015612024.1 | 3 - 4 | 677 | 85 | |
XM_015756539.2 | XP_015612025.1 | 3 - 4 | 678 | 85 | ||
GenBank mRNA | AK103074.1 | - | 3 - 4 | 652 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
