Assay Details
| Gene Symbol: LOC4347697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756538.2 | XP_015612024.1 | 6 - 7 | 965 | 136 | |
| XM_015756539.2 | XP_015612025.1 | 6 - 7 | 966 | 136 | ||
| GenBank mRNA | AK103074.1 | - | 6 - 7 | 940 | 136 | |
More Information
Set Membership: |
|

LOC4347697
Genomic Map