Assay Details
| Gene Symbol: LOC4348805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759021.2 | XP_015614507.1 | 19 - 20 | 3356 | 97 | |
| XM_015759022.2 | XP_015614508.1 | 19 - 20 | 3369 | 97 | ||
| XM_015759023.1 | XP_015614509.1 | 19 - 20 | 3346 | 97 | ||
| XM_015759024.2 | XP_015614510.1 | 19 - 20 | 3359 | 97 | ||
| GenBank mRNA | AK068079.1 | - | 19 - 20 | 3347 | 97 | |
More Information
Set Membership: |
|

LOC4348805
Genomic Map