Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 15 - 16 | 1489 | 65 | |
| XM_015758827.1 | XP_015614313.1 | 11 - 12 | 954 | 65 | ||
| GenBank mRNA | AK318583.1 | - | 15 - 16 | 1588 | 65 | |
More Information
Set Membership: |
|

LOC9270457
Genomic Map