Assay Details
| Gene Symbol: LOC9270457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758826.1 | XP_015614312.1 | 5 - 6 | 545 | 89 | |
| XM_015758827.1 | XP_015614313.1 | 1 - 2 | 66 | 89 | ||
| GenBank mRNA | AK318583.1 | - | 5 - 6 | 644 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015758827.1 transcripts.
Set Membership: |
|

LOC9270457
Genomic Map