Assay Details
| Gene Symbol: LOC4349917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761984.2 | XP_015617470.1 | 13 - 14 | 1339 | 137 | |
| XM_015761985.2 | XP_015617471.1 | 12 - 13 | 1249 | 137 | ||
| GenBank mRNA | AK064320.1 | - | 13 - 14 | 1315 | 137 | |
| AK101592.1 | - | 12 - 13 | 1244 | 137 | ||
More Information
Set Membership: |
|

LOC4349917
Genomic Map