Assay Details
Gene Symbol: LOC4330328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768461.2 | XP_015623947.1 | 3 - 4 | 553 | 100 | |
GenBank mRNA | AK069103.1 | - | 3 - 4 | 461 | 100 | |
FJ666318.1 | - | 3 - 4 | 460 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
