Assay Details
Gene Symbol: LOC4330328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768461.2 | XP_015623947.1 | 4 - 5 | 649 | 73 | |
GenBank mRNA | AK069103.1 | - | 4 - 5 | 557 | 73 | |
FJ666318.1 | - | 4 - 5 | 556 | 73 | ||
U78782.1 | - | 4 - 5 | 457 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
