Assay Details
Gene Symbol: LOC4330328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768461.2 | XP_015623947.1 | 5 - 6 | 693 | 78 | |
GenBank mRNA | AK069103.1 | - | 5 - 6 | 601 | 78 | |
FJ666318.1 | - | 5 - 6 | 600 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
