Assay Details
Gene Symbol: LOC4330328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768461.2 | XP_015623947.1 | 7 - 8 | 879 | 64 | |
GenBank mRNA | AK069103.1 | - | 7 - 8 | 787 | 64 | |
FJ666318.1 | - | 7 - 8 | 786 | 64 | ||
U78782.1 | - | 7 - 8 | 687 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
