Assay Details
| Gene Symbol: LOC4330387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770608.2 | XP_015626094.1 | 2 - 3 | 387 | 146 | |
| XM_015770609.2 | XP_015626095.1 | 2 - 3 | 388 | 146 | ||
| GenBank mRNA | AK071581.1 | - | 2 - 3 | 380 | 146 | |
| AK119260.1 | - | 2 - 3 | 328 | 145 | ||
More Information
Set Membership: |
|

LOC4330387
Genomic Map