Assay Details
| Gene Symbol: LOC4330387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770608.2 | XP_015626094.1 | 3 - 4 | 454 | 97 | |
| XM_015770609.2 | XP_015626095.1 | 3 - 4 | 455 | 97 | ||
| GenBank mRNA | AK071581.1 | - | 3 - 4 | 447 | 97 | |
| AK119260.1 | - | 3 - 4 | 395 | 97 | ||
More Information
Set Membership: |
|

LOC4330387
Genomic Map