Assay Details
Gene Symbol: LOC4330787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771634.1 | XP_015627120.1 | 13 - 14 | 2010 | 123 | |
GenBank mRNA | AK064259.1 | - | 13 - 14 | 1992 | 123 | |
AK068256.1 | - | 13 - 14 | 1992 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
