Assay Details
Gene Symbol: LOC4330787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771634.1 | XP_015627120.1 | 3 - 4 | 703 | 73 | |
GenBank mRNA | AK064259.1 | - | 3 - 4 | 687 | 73 | |
AK068256.1 | - | 3 - 4 | 685 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
