Assay Details
Gene Symbol: LOC4331112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771322.1 | XP_015626808.1 | 13 - 14 | 1328 | 85 | |
XM_015771323.2 | XP_015626809.1 | 13 - 14 | 1312 | 85 | ||
GenBank mRNA | AK058270.1 | - | 2 - 3 | 124 | 85 | |
AK100446.1 | - | 13 - 14 | 1221 | 85 | ||
D78136.1 | - | 12 - 13 | 1178 | 85 | ||
D85751.1 | - | 13 - 14 | 1276 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
