Assay Details
| Gene Symbol: LOC4331112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771322.1 | XP_015626808.1 | 15 - 16 | 1536 | 64 | |
| XM_015771323.2 | XP_015626809.1 | 15 - 16 | 1520 | 64 | ||
| GenBank mRNA | AK058270.1 | - | 4 - 5 | 332 | 64 | |
| AK100446.1 | - | 15 - 16 | 1429 | 64 | ||
| D78136.1 | - | 14 - 15 | 1386 | 64 | ||
| D85751.1 | - | 15 - 16 | 1484 | 64 | ||
More Information
Set Membership: |
|

LOC4331112
Genomic Map