Assay Details
| Gene Symbol: LOC4331112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771322.1 | XP_015626808.1 | 1 - 2 | 167 | 63 | |
| GenBank mRNA | AK100446.1 | - | 1 - 2 | 60 | 63 | |
| D85751.1 | - | 1 - 2 | 115 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771322.1 transcripts.
Set Membership: |
|

LOC4331112
Genomic Map