Assay Details
Gene Symbol: LOC4331112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771322.1 | XP_015626808.1 | 8 - 9 | 908 | 105 | |
XM_015771323.2 | XP_015626809.1 | 8 - 9 | 892 | 105 | ||
GenBank mRNA | AK066398.1 | - | 5 - 6 | 482 | 105 | |
AK100446.1 | - | 8 - 9 | 801 | 105 | ||
D78136.1 | - | 7 - 8 | 758 | 105 | ||
D85751.1 | - | 8 - 9 | 856 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
