Assay Details
| Gene Symbol: LOC4331301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776752.2 | XP_015632238.1 | 1 | 567 | 83 | |
| GenBank mRNA | AK103618.1 | - | 1 | 556 | 83 | |
| AY466108.1 | - | 1 | 535 | 83 | ||
| BT129391.1 | - | 1 | 556 | 83 | ||
More Information
Set Membership: |
|

LOC4331301
Genomic Map